Human recombinant SP-D (from cells)
Fournisseur: ProSci Inc.
91-491
PRSI91-491EA 549 EUR InStock
PRSI91-491
Human recombinant SP-D (from cells)
Protéines et Peptides
Surfactant Pulmonary-Associated Protein D (SP-D) is a 43 kDa member of the collectin family of innate immune modulators. Its principal components consist of a collagen-like region and a C-terminal carbohydrate recognition domain (CRD), a structure that places it in a subset of pattern recognition proteins termed defense collagens. SP-D is constitutively secreted by alveolar lining cells and epithelium associated with tubular structures and induced in cardiac smooth muscle and endothelial cells. It binds both secreted and transmembrane proteins that transduce its function. It binds human neutrophil defensins, modulating influenza anti-viral defense. It binds MD-2/LY96, a secreted protein that cooperates with Toll-like receptors (TLRs) in the response of macrophages to bacterial lipopolysaccharides (LPS) or cell wall components. It also binds macrophage CD14 and TLRs directly, blocking binding of LPS and down-regulating TNF- alpha secretion. SP-D binding of both SIRP alpha and the calreticulin/CD91 complex on macrophages allows for a graded response to environmental challenge.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité