Human recombinant Serine Protease Inhibitor Kazal-Type 1 (from cells)
Fournisseur: ProSci Inc.
91-492
PRSI91-492EA 654 EUR InStock
PRSI91-492
Human recombinant Serine Protease Inhibitor Kazal-Type 1 (from cells)
Protéines et Peptides
Serine Protease Inhibitor Kazal-Type 1 (SPINK1) is a trypsin inhibitor that prevent the trypsin-catalysed premature activation of zymogens within the pancreas. Defects in SPINK1 are a cause of pancreatitis (PCTT). A disease characterised by the presence of calculi in pancreatic ducts. It causes severe abdominal pain attacks. Defects in SPINK1 are the cause of susceptibility to tropical calcific pancreatitis (TCP). Recombinant SPINK1 protein (rSPINK1) stimulated cell proliferation in benign RWPE as well as cancerous prostate cells. The research result indicated that the potential of SPINK1 as an extracellular therapeutic target in prostate cancer. In contrast, knockdown of SPINK1 in 22RV1 cells inhibited cell proliferation, cell invasion, and tumour growth in xenograft assays.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité