Human recombinant vanin-1 (from cells)
Fournisseur: ProSci Inc.
91-497
PRSI91-497EA 654 EUR InStock
PRSI91-497
Human recombinant vanin-1 (from cells)
Protéines et Peptides
Vanin-1 is a cell membrane protein which contains one CN hydrolase domain and belongs to the CN hydrolase family and BTD/VNN subfamily. Vanin-1 is also a member of the Vanin family of proteins, which share extensive sequence similarity with each other, and also with biotinidase. The family includes secreted and membrane-associated proteins, a few of which have been reported to participate in hematopoietic cell trafficking. Vanin-1 is widely expressed with higher expression in spleen, kidney and blood and overexpressed in lesional psoriatic skin. No biotinidase activity has been demonstrated for any of the vanin proteins; however, they possess pantetheinase activity, which may play a role in oxidative-stress response. Vanin-1 is an epithelial pantetheinase that provides cysteamine to tissue and regulates response to stress. Vanin-1 is expressed by enterocytes, and its absence limits intestinal epithelial cell production of proinflammatory signals. Vanin-1 regulates late adhesion steps of thymus homing under physiological, noninflammatory conditions. The early impact of vanin-1 deficiency on tumor induction was directly correlated to the amount of inflammation and subsequent epithelial proliferation rather than cell death rate. Vanin-1 molecule was shown to be involved in the control of thymus reconstitution following sub-lethal irradiation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité