Human recombinant VSIG2 (from cells)
Fournisseur: ProSci Inc.
91-498
PRSI91-498EA 549 EUR InStock
PRSI91-498
Human recombinant VSIG2 (from cells)
Protéines et Peptides
V-Set and Immunoglobulin Domain-Containing Protein 2 (VSIG2) is presumably a 50-60 kDa single-pass type I transmembrane (glyco)protein which contains one Ig-like C2-type (immunoglobulin-like) domain and one Ig-like V-type (immunoglobulin-like) domain. VSIG2 is highly expressed in the stomach, colon, prostate, trachea and thyroid glands and weakly in bladder and lung. V-set domains are Ig-like domains resembling the antibody variable domain. V-set domains are found in diverse protein families, including immunoglobulin light and heavy chains, in several T-cell receptors such as CD2 (Cluster of Differentiation 2), CD4, CD80, and CD86, in myelin membrane adhesion molecules, in junction adhesion molecules (JAM), in tyrosine-protein kinase receptors, and in the programmed cell death protein 1 (PD1). It shows expression in stomach and prostate by Northern blot, and likely participates in cell adhesion. Human VSIG2 precursor is 327 amino acids in length.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité