Human recombinant CLM9 (from cells)
Fournisseur: ProSci Inc.
91-515
PRSI91-515EA 654 EUR InStock
PRSI91-515
Human recombinant CLM9 (from cells)
Protéines et Peptides
CMRF35-Like Molecule 9 (CD300LG) is a single-pass type I membrane protein which belongs to the CD300 family. CD300LG has one Ig-like V-type domain which mediates binding to lymphocyte. CD300LG is highly expressed in heart, skeletal muscle and placenta. CD300LG acts as a receptor which may mediate L-selectin-dependent lymphocyte rollings. CD300LG also binds SELL in a calcium dependent manner and lymphocyte. CD300LG may play a important role in molecular traffic across the capillary endothelium.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité