Human recombinant CD5L (from cells)
Fournisseur: ProSci Inc.
91-518
PRSI91-518EA 549 EUR InStock
PRSI91-518
Human recombinant CD5L (from cells)
Protéines et Peptides
CD5 Antigen-Like (CD5L) is a soluble protein that belongs to group B of the scavenger receptor cysteine-rich (SRCR) superfamily and contains three SRCR domains. CD5L is a secreted glycoprotein and expressed by macrophages presentin lymphoid tissues. It binds to myelomonocytic and lymphoid cells and may play an important role in the regulation of the innate and adaptive immune systems. CD5L functions as a pattern recognition molecule by binding both lipoteichoic acid (LTA) on Gram positive and lipopolysaccharide (LPS) on Gram-negative bacteria and the SRCR domain one of CD5L retains both the LPS and LTA binding activities. Furthermore, CD5L seems to play a role as an inhibitor of apoptosis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité