Human recombinant Cerberus 1 (from cells)
Fournisseur: ProSci Inc.
91-521
PRSI91-521EA 773 EUR InStock
PRSI91-521
Human recombinant Cerberus 1 (from cells)
Protéines et Peptides
Cerberus 1 is a secreted glycoprotein that forms disulfide-linked homodimers. It is a cytokine member of the DAN domain family of BMP antagonists that includes DAN (DAND1), Gremlin/Drm (DAND2), PRDC (Protein Related to Dan and Cerberus, DAND3), and COCO/Dante (DAND5). DAN family members contain a cysteine knot domain that is homologous to that found in other TGF-beta superfamily ligands. At the onset of gastrulation, Cerberus 1 is transiently expressed in anterior endodermal structures in response to Nodal and Shh. Cerberus 1 binds BMP-4 and Nodal and inhibits their activities. The inhibitory functions of Cerberus favor mesodermal development in the anterior region of the gastrula and suppresses posterior mesodermal differentiation. In chick and Xenopus, Cerberus 1 also regulates, but is not required for embryonic left-right polarization, neurulation, and head and heart induction.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité