Human recombinant Corneodesmosin (from cells)
Fournisseur: ProSci Inc.
91-526
PRSI91-526EA 471 EUR InStock
PRSI91-526
Human recombinant Corneodesmosin (from cells)
Protéines et Peptides
Corneodesmosin is a protein that is encoded by the CDSN gene in humans. This gene encodes a protein found in corneodesmosomes, which localize to human epidermis and other cornified squamous epithelia. During maturation of the cornified layers, the protein undergoes a series of cleavages, which are thought to be required for desquamation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité