Human recombinant Carboxypeptidase A1 (from Cells)
Fournisseur: ProSci Inc.
91-528
PRSI91-528EA 789 EUR InStock
PRSI91-528
Human recombinant Carboxypeptidase A1 (from Cells)
Enzymes
Carboxypeptidase A1 (CPA1) is secreted as a pancreatic peptidase that comes from the precursor form of inactive procarboxypeptidase. CPA1 comprises a signal peptide, a pro region and a mature chain, and can be activated after cleavage of the pro peptide. It has a free C-terminal carboxyl group, with the preference of residues with aromatic or branched aliphatic side chains. CPA1 cleaves the C-terminal amide or ester bond of peptides and involves in zymogen inhibition. Three different forms of human pancreatic procarboxypeptidase A have been isolated. In contrast to procarboxypeptidase B which was always secreted by the pancreas as a monomer, procarboxypeptidase A occurs as a monomer and/or associated to one or two functionally different proteins, such as zymogen E.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité