Human recombinant CXCL1 (from cells)
Fournisseur: ProSci Inc.
91-532
PRSI91-532EA 734 EUR InStock
PRSI91-532
Human recombinant CXCL1 (from cells)
Protéines et Peptides
Chemokine (C-X-C motif) Ligand 1 Protein (CXCL1) is a growth factor for melanoma cells and a chemotaxin for neutrophils and a member of the CXC chemokine family that is a potent neutrophil attractant and activator and is also active toward basophils. CXCL1 is expressed by macrophages, neutrophils and epithelial cells; it has neutrophil chemoattractant activity. CXCL1 plays a critical nonredundant role in the development of experimental Lyme arthritis and carditis via CXCR2-mediated recruitment of neutrophils into the site of infection and may also have important pro-nociceptive effects via its direct actions on sensory neurons, and may induce long-term changes that involve protein synthesis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité