Human recombinant EDIL3 (from cells)
Fournisseur: ProSci Inc.
91-534
PRSI91-534EA 549 EUR InStock
PRSI91-534
Human recombinant EDIL3 (from cells)
Protéines et Peptides
EGF-Like Repeat and Discoidin I-Like Domain-Containing Protein 3 (EDIL3) is a 52 kDa extracellular matrix protein that is expressed by endothelial tissues during embryonic vascular development. EDIL3 becomes quiescent at the time of birth, and is no longer expressed in normal adult tissues. EDIL3 has been found to be re-expressed in a number of human tumors as well as in ischemic muscles and ischemic brain tissue, which may play an important role in adult angiogenesis. EDIL3 promotes adherence and migration of endothelial cells, and acts as an endothelial cell survival agent through upregulation of Bcl-2 expression. EDIL3 has also been shown to be an endogenous inhibitor of inflammatory cell recruitment by interfering with the integrin LFA-1-dependent leukocyte-endothelial adhesion. Human EDIL3 is synthesized as a precursor with a 16 amino acid signal sequence and a 464 amino acid mature chain.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité