Human Recombinant GLIPR1 (from Cells)
Fournisseur: ProSci Inc.
91-536
PRSI91-536EA 852 EUR InStock
PRSI91-536
Human Recombinant GLIPR1 (from Cells)
Protéines et Peptides
Glioma Pathogenesis-Related Protein 1 (GLIPR1) is a member of the CRISP family. It is a single-pass membrane protein with 266 amino acids. GLIPR1 is highly expressed in the lung and kidney and lowly expressed in the heart and liver. It is also highly expressed in cell lines that are derived from nervous system tumors arising from glia; conversely, it is lowly expressed in non-glial-derived nervous system tumor cell lines. GLIPR1 is only expressed in brain tumor glioblastoma multiforme/astrocytoma and not expressed in other nervous system tumors nor normal adult or fetal tissues.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité