Human recombinant LRRTM2 (from cells)
Fournisseur: ProSci Inc.
91-538
PRSI91-538EA 654 EUR InStock
PRSI91-538
Human recombinant LRRTM2 (from cells)
Protéines et Peptides
Leucine-Rich Repeat Transmembrane Neuronal Protein 2 (LRRTM2) is a single-pass type I membrane protein that belongs to the LRRTM family. It contains ten LRR (leucine-rich) repeats, one LRRCT domain, and one LRRNT domain. LRRTM2 is expressed in neuronal tissues, and it interacts with DLG4 and NRXN1. LRRTM2 has been suggested to be involved in the development and maintenance of excitatory synapses in the vertebrate nervous system. LRRTM2 also regulates the surface expression of AMPA receptors. LRRTM2 acts as a ligand for the presynaptic receptors NRXN1-A and NRXN1-B.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité