Human recombinant LYVE1 (from cells)
Fournisseur: ProSci Inc.
91-539
PRSI91-539EA 734 EUR InStock
PRSI91-539
Human recombinant LYVE1 (from cells)
Protéines et Peptides
Lymphatic Vessel Endothelial Hyaluronic Acid Receptor 1 is a single-pass type I membrane protein. LYVE-1 is a CD44 homolog found primarily on lymphatic endothelial cells 1. LYVE-1 mainly expressed in endothelial cells lining lymphatic vessels. While LYVE-1 functions is a Ligand-specific transporter trafficking between intracellular organelles (TGN) and the plasma membrane. LYVE-1 plays a role in autocrine regulation of cell growth mediated by growth regulators containing cell surface retention sequence binding (CRS). It may act as an hyaluronan (HA) transporter, either mediating its uptake for catabolism within lymphatic endothelial cells themselves, or its transport into the lumen of afferent lymphatic vessels for subsequent re-uptake and degradation in lymph nodes.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité