Human recombinant protocadherin 10 (from cells)
Fournisseur: ProSci Inc.
91-543
PRSI91-543EA 525 EUR InStock
PRSI91-543
Human recombinant protocadherin 10 (from cells)
Protéines et Peptides
Protocadherin-10 (PCDH10) is a single-pass type I membrane protein that contains six extracellular cadherin domains, one transmembrane domain and one cytoplasmic tail differing from those of the classical cadherins. As a potential calcium-dependent cell-adhesion neuronal receptor, it may plays a role in the establishment and function of specific cell-cell connections in the brain. PCDH10 moderately expressed in all regions of the brain examined, as well as in testis and ovary, and low expression in all other tested tissues.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité