Human recombinant NGR (from cells)
Fournisseur: ProSci Inc.
91-545
PRSI91-545EA 528 EUR InStock
PRSI91-545
Human recombinant NGR (from cells)
Protéines et Peptides
Nogo Receptor (NgR) is a glycosylphosphoinositol (GPI)-anchored protein that belongs to the Nogo recptor family. Human NgR is predominantly expressed in neurons and their axons in the central nervous systems. As a receptor for myelin-derived proteins Nogo, myelin-associated glycoprotein (MAG) and myelin oligodendrocyte glycoprotein (OMG), NgR mediates axonal growth inhibition and may play a role in regulating axonal regeneration and plasticity in the adult central nervous system. NgR may be proposed as a potential drug target for treatment of various neurological conditions. Additionally, NgR may play a role in regulating the function of gap junctions.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité