Human recombinant syndecan 2 (from cells)
Fournisseur: ProSci Inc.
91-547
PRSI91-547EA 490 EUR InStock
PRSI91-547
Human recombinant syndecan 2 (from cells)
Protéines et Peptides
Syndecan-2 is a member of the Syndecans family comprised of type I transmembrane heparan sulfate proteoglycans (HSPG) that are involved in the regulation of many cellular processes. Four sub-types of mammalian Syndecans have been reported and among them. Syndecan-2 plays a role in the cancer development. It can affect the basal and chemotherapy-induced apoptosis in osteosarcoma. It can also suppress MMP2 activation, suppressing metastasis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité