Human recombinant Semenogelin-1 (from cells)
Fournisseur: ProSci Inc.
91-549
PRSI91-549EA 654 EUR InStock
PRSI91-549
Human recombinant Semenogelin-1 (from cells)
Protéines et Peptides
Semenogelin-1 (SEMG1) is the predominant protein in semen; it is a secretory protein involved in the formation of a gel matrix entrapping the accessory gland secretions and ejaculated spermatozoa. The prostate-specific antigen (PSA) protease processes SEMG1 into smaller peptides, each possibly having a separate function. In the proteolysis process, Alpha-inhibin-92 and alpha-inhibin-31 are produced; they inhibit the secretion of pituitary follicle-stimulating hormone. At the same time, it breaks down the gel matrix, allowing the spermatozoa to move more freely.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité