Human Recombinant HAI-2 (from Cells)
Fournisseur: ProSci Inc.
91-552
PRSI91-552EA 584 EUR InStock
PRSI91-552
Human Recombinant HAI-2 (from Cells)
Protéines et Peptides
Hepatocyte Growth Factor Activator Inhibitor Type 2 (HAI2) is a single-pass type I membrane protein and contains two BPTI/Kunitz inhibitor domains. The first Kunitz domain is mainly responsible for the inhibitory activity against hepatocyte growth factor activator (HGFA). HAI2 is expressed in placenta, kidney, pancreas, prostate, testis, thymus and trachea. HAI2 serves as a inhibitor of HGF activator. It also inhibits plasmin, plasma and tissue kallikrein and factor XIa. Defects in HAI2 are the cause of diarrhea type 3 (DIAR3), also known as congenital sodium diarrhea (CSD).
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité