Human recombinant THSD1 (from cells)
Fournisseur: ProSci Inc.
91-553
PRSI91-553EA 377 EUR InStock
PRSI91-553
Human recombinant THSD1 (from cells)
Protéines et Peptides
Thrombospondin Type-1 Domain-Containing Protein 1 (THSD1) is a single-pass type I membrane protein. THSD1 contains a signal peptide and one TSP type-1 domain that is found in thrombospondin. THSD1 is a good novel candidate for TSG as it has been mapped to 13q14. Alternatively spliced transcript variants encoding distinct isoforms have been observed. THSD1 may be involved in the complement pathway.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité