Human recombinant vanin-2 (from cells)
Fournisseur: ProSci Inc.
91-555
PRSI91-555EA 654 EUR InStock
PRSI91-555
Human recombinant vanin-2 (from cells)
Protéines et Peptides
Vascular Non-Inflammatory Molecule 2 (VNN2) is a member of the CN hydrolase family. The family includes secreted and membrane-associated proteins, a few of which have been reported to participate in hematopoietic cell trafficking. they possess pantetheinase activity, which may play a role in oxidative-stress response. VNN2 is a GPI-anchored cell surface molecule that plays a role in transendothelial migration of neutrophils. VNN2 involved in the thymus homing of bone marrow cells. In addition, VNN2 may regulate beta-2 integrin-mediated cell adhesion, migration and motility of neutrophil.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité