Human recombinant ZG16 (from cells)
Fournisseur: ProSci Inc.
91-556
PRSI91-556EA 760 EUR InStock
PRSI91-556
Human recombinant ZG16 (from cells)
Protéines et Peptides
Zymogen Granule Membrane Protein 16 (ZG16) belongs to the jacalin lectin family. ZG16 is highly expressed in liver and is detected at lower levels in colon, ileum and jejunum. ZG16 may play a role in protein trafficking. In addition, ZG16 may act as a linker molecule between the submembranous matrix on the luminal side of zymogen granule membrane (ZGM) and aggregated secretory proteins during granule formation in the TGN.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité