Human recombinant MASP1 (from cells)
Fournisseur: ProSci Inc.
91-557
PRSI91-557EA 884 EUR InStock
PRSI91-557
Human recombinant MASP1 (from cells)
Protéines et Peptides
Mannan-Binding Lectin Serine Protease 1 (MASP-1) belongs to the peptidase S1 family. MASP1 contains two CUB domains, one EGF-like domain, one peptidase S1 domain and two Sushi (CCP/SCR) domains. MASP1 is primarily expressed in liver. MASP1 involved in the lectin pathway of the complement, performs a key role in innate immunity by recognizing pathogens through patterns of sugar moieties and neutralizing them. MASP1 is synthesized as a zymogen and activated when it complexes with the pathogen recognition molecules of lectin pathway, the mannose-binding lectin and the ficolins. MASP1 is not directly involved in complement activation but may act as an amplifier of complement activation by cleaving complement C2 or by activating another complement serine protease, MASP2. MASP1 is also able to cleave fibrinogen and factor XIII and may may be involved in coagulation. MASP1 is inhibited by SERPING1 and A2M.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité