Human recombinant LIPF (from cells)
Fournisseur: ProSci Inc.
91-558
PRSI91-558EA 884 EUR InStock
PRSI91-558
Human recombinant LIPF (from cells)
Protéines et Peptides
Gastric Triacylglycerol Lipase (LIPF) belongs to the AB hydrolase superfamily. LIPF is an important lipase during the digestion of dietary lipids in cystic fibrosis. LIPF is involved in the digestion of dietary triglycerides in the gastrointestinal tract, and responsible for 30% of fat digestion processes occurring in human. LIPF is secreted by gastric chief cells in the fundic mucosa of the stomach, and it hydrolyzes the ester bonds of triglycerides under acidic pH conditions. LIPF acts distinct roles in neutral lipid metabolism.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité