Human recombinant IFN alpha 6 (from cells)
Fournisseur: ProSci Inc.
91-559
PRSI91-559EA 852 EUR InStock
PRSI91-559
Human recombinant IFN alpha 6 (from cells)
Protéines et Peptides
Interferon alpha-6 (IFN- alpha6) is a secreted protein which belongs to the alpha/ beta interferon family. IFN- alpha6 is produced by macrophages, expressed at low level, only 1.0% of the average gene in this release. IFN- alpha6 contains interferon alpha, beta and delta domain. IFN- alpha has antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité