Human recombinant POMGNT1 (from cells)
Fournisseur: ProSci Inc.
91-563
PRSI91-563EA 789 EUR InStock
PRSI91-563
Human recombinant POMGNT1 (from cells)
Protéines et Peptides
Protein O-Linked-Mannose beta -1 2-N-Acetylglucosaminyltransferase 1 (POMGNT1) belongs to the Glycosyltransferase 13 family. Amino acid residues between 299-311 are important for both protein expression and enzymatic activity. The minimal catalytic domain is located between positions 299-651. It is suggested that the stem domain of the soluble form is unnecessary for activity, but that some amino acids play a crucial role in the membrane-bound form. Defects in POMGNT1 are the cause of muscular dystrophy-dystroglycanopathy congenital with brain and eye anomalies type A3 (MDDGA3).
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité