Human recombinant TXNDC12 (from cells)
Fournisseur: ProSci Inc.
91-564
PRSI91-564EA 884 EUR InStock
PRSI91-564
Human recombinant TXNDC12 (from cells)
Protéines et Peptides
Thioredoxin Domain-Containing Protein 12 belongs to the thioredoxin superfamily. In this family, proteins possess a thioredoxin fold with a consensus active-site sequence (CxxC) and have roles in redox regulation, defense against oxidative stress, refolding of disulfide-containing proteins, and regulation of transcription factors. TXNDC12 is widely expressed in many tissues and contains one thioredoxin domain.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité