Human recombinant SOD2 (from E. coli)
Fournisseur: ProSci Inc.
91-566
PRSI91-566EA 884 EUR InStock
PRSI91-566
Human recombinant SOD2 (from E. coli)
Protéines et Peptides
Superoxide Dismutase (SOD2) is a number of the iron/manganese superoxide dismutase family. SOD2 is a mitochondrial protein that forms a homotetramer and binds one manganese ion per subunit. The SOD2 protein transforms toxic superoxide and a byproduct of the mitochondrial electron transport chain into hydrogen peroxide and diatomic oxygen. Genetic variation in SOD2 is associated with microvascular complications of diabetes type 6 (MVCD6), idiopathic cardiomyopathy (IDC), sporadic motor neuron disease, and cancer. SOD2 destroys superoxide anion radicals which are usually produced within the cells and which are toxic to biological systems.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité