Human recombinant ITM2B (from cells)
Fournisseur: ProSci Inc.
91-567
PRSI91-567EA 761 EUR InStock
PRSI91-567
Human recombinant ITM2B (from cells)
Protéines et Peptides
Integral Membrane Protein 2B (ITM2B) is expressed in the Golgi and on the cell surface. ITM2B forms homodimer through disulfide-linked interaction with SPPL2A, SPPL2B and APP. ITM2B is expressed in brain and the other tissues. Defects in ITM2B cause cerebral amyloid angiopathy ITM2B-related type 1(CAA-ITM2B1) and amyloid angiopathy ITM2B-related type 2(CAA-ITM2B2). CAA-ITM2B1 is characterized by amyloid deposition in the walls of cerebral blood vessels and neurodegeneration in the central nervous system. CAA-ITM2B2 characterized by amyloid deposition in the walls of the blood vessels of the cerebrum, choroid plexus, cerebellum, spinal cord and retina.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité