Human recombinant Pancreatic Lipase-Related 1 (from cells)
Fournisseur: ProSci Inc.
91-568
PRSI91-568EA 584 EUR InStock
PRSI91-568
Human recombinant Pancreatic Lipase-Related 1 (from cells)
Enzymes
Pancreatic Lipase-Related Protein 1 (PNLIPRP1) belongs to the Lipase family within the AB hydrolase superfamily. PNLIPRP1 is a secreted protein and contains one PLAT domain. PNLIPRP1 is involved in lipid metabolic process, acting as a negative regulator of pancreatic lipase activity by competing with pancreatic lipase for colipase occupancy. PNLIPRP1 may play a role in inhibiting dietary triglyceride digestion, but it lacks detectable lipase activity towards triglycerides, diglycerides, phosphatidylcholine, galactolipids or cholesterol esters.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité