Human recombinant SPESP1 (from cells)
Fournisseur: ProSci Inc.
91-570
PRSI91-570EA 852 EUR InStock
PRSI91-570
Human recombinant SPESP1 (from cells)
Protéines et Peptides
Sperm Equatorial Segment Protein 1 (SPESP1) is a member of the SPESP1 family. SPESP1 is highly expressed in the testis, where it is localized to the acrosome of postmeiotic stages of spermiogenesis; it is expressed at lower levels in the placenta and fetal lung. SPESP1 is involved in the multicellular organisimal development. Disruption of SPESP1 leads to abnormal distribution of sperm proteins resulting in a detached membrane from the equatorial segment and less fertile sperm. SPESP1 may interact with IZUMO1 and MN9 antigen and it contains an N-glycosylation site as well as several cAMP-dependent kinase, protein kinase C, and casein kinase II consensus phosphorylation sites.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité