Human recombinant Dickkopf-related 1 (from cells)
Fournisseur: ProSci Inc.
91-573
PRSI91-573EA 852 EUR InStock
PRSI91-573
Human recombinant Dickkopf-related 1 (from cells)
Protéines et Peptides
Dickkopf-related protein 1(DKK-1), is a member of the dickkopf family. DKK1 secreted proteins with two cysteine-rich domains separated by a linker region. It antagonizes canonical Wnt signaling by inhibiting LRP5/6 interaction with Wnt and by forming a ternary complex with the transmembrane protein KREMEN that promotes internalization of LRP5/6. DKKs play an important role in vertebrate development, where they locally inhibit Wnt regulated processes such as antero-posterior axial patterning, limb development, somitogenesis and eye formation. In the adult, Dkks are implicated in bone formation and bone disease, cancer and Alzheimer disease.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-8 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité