Human recombinant TNF Receptor I (from E. coli)
Fournisseur: ProSci Inc.
91-574
PRSI91-574EA 654 EUR InStock
PRSI91-574
Human recombinant TNF Receptor I (from E. coli)
Protéines et Peptides
Tumor necrosis factor receptor superfamily member 1A (Tnfrsf1a) is a member of the tumor necrosis factor receptor superfamily. Tnfrsf1a is one of the major receptors for the tumor necrosis factor-alpha. It can activate the transcription factor NF-κB, mediate apoptosis, and function as a regulator of inflammation. Antiapoptotic protein BCL2-associated athanogene 4 (BAG4/SODD) and adaptor proteins TRADD and TRAF2 have been shown to interact with this receptor, and thus play regulatory roles in the signal transduction mediated by the receptor. Germline mutations of the extracellular domains of this receptor were found to be associated with the human genetic disorder called tumor necrosis factor associated periodic syndrome (TRAPS) or periodic fever syndrome
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité