Mouse recombinant LIF (from E. coli)
Fournisseur: ProSci Inc.
91-575
PRSI91-575EA 525 EUR InStock
PRSI91-575
Mouse recombinant LIF (from E. coli)
Protéines et Peptides
Mouse Leukemia inhibitory factor(lif)is a secreted protein which belongs to the LIF/OSM family.LIF has been implicated in a many physiological processes including development, hematopoiesis, bone metabolism, and inflammation. it has the capacity to induce terminal differentiation in leukemic cells. Its activities include the induction of hematopoietic differentiation in normal and myeloid leukemia cells, the induction of neuronal cell differentiation, and the stimulation of acute-phase protein synthesis in hepatocytes.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité