Human recombinant FGF2 (from E. coli)
Fournisseur: ProSci Inc.
91-594
PRSI91-594EA 654 EUR InStock
PRSI91-594
Human recombinant FGF2 (from E. coli)
Protéines et Peptides
Fibroblast growth factor 2(FGF2) is a secreted protein and belongs to the heparin-binding growth factors family. FGF2 is produced by epithelial, tumor and other cell types. It involved in developmental processes and regulates differentiation, proliferation, and migration, FGF2 is a critical factor for growing embryonic stem cells in culture without inducing differentiation. FGF2 has a high affinity for heparan sulfate and binding is a step in the FGF basic activation of FGFR tyrosine kinase.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité