Mouse recombinant CADM1
Fournisseur: ProSci Inc.
91-601
PRSI91-601EA 588 EUR InStock
PRSI91-601
Mouse recombinant CADM1
Protéines et Peptides
Cell adhesion molecule 1(CADM1) is a single-pass type I membrane protein and belongs to the nectin family. It contains 2 Ig-like C2-type (immunoglobulin-like) domains and 1 Ig-like V-type (immunoglobulin-like) domain. CADM1 acts as a tumor suppressor in non-small-cell lung cancer (NSCLC) cells. Interaction with CRTAM promotes natural killer (NK) cell cytotoxicity and interferon-gamma (IFN-gamma) secretion by CD8+ cells in vitro as well as NK cell-mediated rejection of tumors expressing CADM3 in vivo. CADM1 may contribute to the less invasive phenotypes of lepidic growth tumor cells. In mast cells, it may mediate attachment to and promote communication with nerves. CADM1, together with MITF, is essential for development and survival of mast cells in vivo. The protein acts as a synaptic cell adhesion molecule and plays a role in the formation of dendritic spines and in synapse assembly. It may be involved in neuronal migration, axon growth, pathfinding, and fasciculation on the axons of differentiating neurons. CADM1 may play diverse roles in the spermatogenesis including in the adhesion of spermatocytes and spermatids to Sertoli cells and for their normal differentiation into mature spermatozoa.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité