Human recombinant TDGF1 (from cells)
Fournisseur: ProSci Inc.
91-604
PRSI91-604EA 947 EUR InStock
PRSI91-604
Human recombinant TDGF1 (from cells)
Protéines et Peptides
Teratocarcinoma-derived growth factor 1(TDGF1) is a Cell membrane protein and contains 1 EGF-like domain. The protein plays an essential role in embryonic development and tumor growth. Mutations in this gene are associated with forebrain defects. It also may play a role in the determination of the epiblastic cells that subsequently give rise to the mesoderm.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité