Mouse recombinant Granzyme B
Fournisseur: ProSci Inc.
91-607
PRSI91-607EA 789 EUR InStock
PRSI91-607
Mouse recombinant Granzyme B
Protéines et Peptides
Granzyme B(GZMB) contains 1 peptidase S1 domain and belongs to the peptidase S1 family. This enzyme is necessary for target cell lysis in cell-mediated immune responses. It cleaves after Asp and seems to be linked to an activation cascade of caspases (aspartate-specific cysteine proteases) responsible for apoptosis execution. The protein cleaves caspase-3, -7, -9 and 10 to give rise to active enzymes mediating apoptosis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité