Human recombinant PTH1 receptor (from cells)
Fournisseur: ProSci Inc.
91-608
PRSI91-608EA 588 EUR InStock
PRSI91-608
Human recombinant PTH1 receptor (from cells)
Protéines et Peptides
Parathyroid hormone 1 receptor(PTH1R) is a multi-pass membrane protein. The protein is expressed in high levels in bone and kidney and regulates calcium ionhomeostasis through activation of adenylate cyclase and phospholipase C. In bone, it is expressed on the surface of osteoblasts. When the receptor is activated through PTH binding, osteoblasts express RANKL (Receptor Activator of Nuclear Factor kB Ligand), which binds to RANK (Receptor Activator of Nuclear Factor kB) on osteoclasts. This turns on osteoclasts to ultimately increase the resorption rate.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité