Human recombinant SLAMF8 (from cells)
Fournisseur: ProSci Inc.
91-613
PRSI91-613EA 525 EUR InStock
PRSI91-613
Human recombinant SLAMF8 (from cells)
Protéines et Peptides
SLAM family member 8(SLAMF8) is a single-pass type I membrane protein and contains 1 Ig-like C2-type (immunoglobulin-like) domain. SLAMF8 encodes a member of the CD2 family of cell surface proteins involved in lymphocyte activation. These proteins are characterized by Ig domains. The protein is expressed in lymphoid tissues, and studies of a similar protein in mouse suggest that it may function during B cell lineage commitment.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité