Human recombinant PRL2 (from E. coli)
Fournisseur: ProSci Inc.
91-619
PRSI91-619EA 588 EUR InStock
PRSI91-619
Human recombinant PRL2 (from E. coli)
Protéines et Peptides
PTP4A2, also known as PRL2 or PTPCAAX2, is short for Protein tyrosine phosphatase type IVA 2. This protein exists in cell membrane, cytoplasm,endosome and membrane. PTP4A2 is often farnesylated during post-translational modification. Farnesylation is required for membrane targeting and for interaction with RABGGTB. The unfarnesylated forms are redirected to the nucleus and cytosol. It can stimulate progression from G1 into S phase during mitosis and promotes tumors. It also inhibits geranylgeranyl transferase type II activity by blocking the association between RABGGTA and RABGGTB.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité