Mouse recombinant carboxypeptidase M
Fournisseur: ProSci Inc.
91-622
PRSI91-622EA 343 EUR InStock
PRSI91-622
Mouse recombinant carboxypeptidase M
Enzymes
Carboxypeptidase M (CPM) belongs to the peptidase M14 family, and exists in cell membrane. The protein binds 1 zinc ion per subunit, and cleavage of C-terminal arginine or lysine residues from polypeptides. CPM specifically removes C-terminal basic residues (Arg or Lys) from peptides and proteins. It is believed to play important roles in the control of peptide hormone and growth factor activity at the cell surface, and in the membrane-localised degradation of extracellular proteins.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité