Mouse recombinant CXCL1
Fournisseur: ProSci Inc.
91-628
PRSI91-628EA 654 EUR InStock
PRSI91-628
Mouse recombinant CXCL1
Protéines et Peptides
Cxcl1, also called Gro, Gro1, Mgsa or Scyb1, is short for growth-regulated alpha protein. The protein belongs to the intercrine alpha (chemokine CxC) family. The N-terminal processed form KC(5-72) of the protein is produced by proteolytic cleavage after secretion from bone marrow stromal cells, and shows a highly enhanced hematopoietic activity. Cxcl1 has chemotactic activity for neutrophils, and contributes to neutrophil activation during inflammation. Hematoregulatory chemokine, in vitro, suppresses hematopoietic progenitor cell proliferation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité