Mouse recombinant SLAMF9
Fournisseur: ProSci Inc.
91-631
PRSI91-631EA 588 EUR InStock
PRSI91-631
Mouse recombinant SLAMF9
Protéines et Peptides
Mouse SLAM family member 9(Slamf9) is a single-pass type I membrane protein. The SLAMF9 gene encodes a member of the signaling lymphocytic activation molecule family. The encoded protein is a cell surface molecule that consists of two extracellular immunoglobulin domains, a transmembrane domain and a short cytoplasmic tail that lacks the signal transduction motifs found in other family members. The slamf9 protein may play a role in the immune response.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité