Human recombinant IL25 (from cells)
Fournisseur: ProSci Inc.
91-634
PRSI91-634EA 654 EUR InStock
PRSI91-634
Human recombinant IL25 (from cells)
Protéines et Peptides
Interleukin 25 (IL-25) belongs to the Interleukin 17 (IL-17) family of proteins, which is comprised of six members (IL-17, IL-17B through IL-17F). These proteins are secreted and are structurally related by sharing a conserved cysteine-knot fold near the C-terminus, but have considerable sequence divergence at the N-terminus. With the exception of IL-17B, which exists as a non-covalently linked dimer, all IL-17 family members are disulfide-linked dimers. IL-17 family proteins are pro-inflammatory cytokines that induce local cytokine production and are involved in the regulation of immune functions. Human interleukin-17E (IL17E), also referred to as Interleukin-25 (IL25), is a distinct member of the IL17 cytokine family comprised of at least six members sharing a conserved cysteine-knot structure but divergent at the N-terminus. IL25 is a glycoprotein secreted as dimers by innate effector eosinophils and basophils, and present at very low levels in various peripheral tissues. IL25, together with IL17B, are ligands for the cytokine receptor IL17BR, and the cross-linking induces NF-κB activation and production of the proinflammatory chemokine IL-8, as well as ERK, JNK, and p38 activation. Overexpression of IL25 gene in transgenic mice suggested that this cytokine can regulate hematopoietic and immune functions, and additionally is identified as a proinflammatory cytokine favoring Th2-type immune responses possibly by enhancing the maintenance and functions of adaptive Th2 memory cells.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité