Mouse recombinant NGF beta (from E. coli)
Fournisseur: ProSci Inc.
91-635
PRSI91-635EA 306 EUR InStock
PRSI91-635
Mouse recombinant NGF beta (from E. coli)
Protéines et Peptides
Mouse beta -NGF is a neurotrophic factor structurally related to BDNF, NT-3 and NT-4. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. beta -NGF is a potent neurotrophic factor that signals through its receptor beta -NGFR, and plays a crucial role in the development and preservation of the sensory and sympathetic nervous systems. beta -NGF also acts as a growth and differentiation factor for B lymphocytes, and enhances B-cell survival.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité