Human recombinant S100 calcium binding A9 (from E. coli)
Fournisseur: ProSci Inc.
91-637
PRSI91-637EA 343 EUR InStock
PRSI91-637
Human recombinant S100 calcium binding A9 (from E. coli)
Protéines et Peptides
Protein S100-A9 (also MRP14 and calgranulin B)is a calcium- and zinc-binding protein which plays a prominent role in the regulation of inflammatory processes and immune response.It can induce neutrophil chemotaxis, adhesion, can increase the bactericidal activity of neutrophils by promoting phagocytosis via activation of SYK, PI3K/AKT, and ERK1/2 and can induce degranulation of neutrophils by a MAPK-dependent mechanism.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité