Human Recombinant GDF11 (from Cells)
Fournisseur: ProSci Inc.
91-641
PRSI91-641EA 947 EUR InStock
PRSI91-641
Human Recombinant GDF11 (from Cells)
Protéines et Peptides
Growth/differentiation factor 11(GDF-11) is a secreted protein, which belongs to the transforming growth factor beta superfamily. GDF-11 controls anterior-posterior patterning by regulating the expression of Hox genes. The secreted signal acts globally to specify positional identity along the anterior/posterior axis during development. GDF11 has been shown to suppress neurogenesis through a pathway similar to that of myostatin, including stopping the progenitor cell-cycle during G-phase. The similarities between GDF11 and myostatin imply a likelihood that the same regulatory mechanisms are used to control tissue size during both muscular and neural development.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité