Human recombinant serpin H1 (from cells)
Fournisseur: ProSci Inc.
91-642
PRSI91-642EA 852 EUR InStock
PRSI91-642
Human recombinant serpin H1 (from cells)
Protéines et Peptides
Serpin H1 is a serine proteinase inhibitors Which belongs to the serpin family. Serpin H1 is induced by heat shock. Serpin H1 localizes to the endoplasmic reticulum lumen and binds specifically to collagen. Thus it is thought to be a molecular chaperone involved in the maturation of collagen molecules. Autoantibodies to this protein have been found in patients with rheumatoid arthritis. Serpin H1 may be a marker for cancer and nucleotide polymorphisms in this gene may be associated with preterm birth caused by preterm premature rupture of membranes.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité