Human recombinant myelin P0 (from cells)
Fournisseur: ProSci Inc.
91-643
PRSI91-643EA 852 EUR InStock
PRSI91-643
Human recombinant myelin P0 (from cells)
Protéines et Peptides
Myelin Protein P0 (MPZ) is a single-pass type I membrane glycoprotein which belongs to the myelin P0 protein family. MPZ contains one Ig-like V-type (immunoglobulin-like) domain, absent in the central nervous system. MPZ is a major component of the myelin sheath in peripheral nerves. It is postulated that MPZ is a structural element in the formation and stabilisation of peripheral nerve myelin, holding its characteristic coil structure together by the interaction of its positively-charged domain with acidic lipids in the cytoplasmic face of the opposed bilayer, and by interaction between hydrophobic globular of adjacent extracellular domains. Defects in MPZ associated with Charcot-Marie-Tooth disease and Dejerine-Sottas disease.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité